missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyclopropyltrifluoroborate, 97%, Thermo Scientific Chemicals

Catalog Number p-4602066
63.66 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1065010-87-8
C3H5BF3K
147.98
CFMLURFHOSOXRC-UHFFFAOYSA-N
potassium cyclopropyltrifluoroborate, potassium cyclopropyltrifluoroboranuide, potassium cyclopropyltrifluororate, potassium cyclopropytrifluoroborate, potassium cycloproyltrifluoroborate, potassiumcyclopropyltrifluoroborate, potassium cyclopropyl-trifluoroborate, cyclopropyltrifluoroborate potassium salt, potassium;cyclopropyl trifluoro boranuide, potassium ion cyclopropyltrifluoroboranuide
23697338
potassium;cyclopropyl(trifluoro)boranuide
[B-](C1CC1)(F)(F)F.[K+]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1065010-87-8
C3H5BF3K
147.98
CFMLURFHOSOXRC-UHFFFAOYSA-N
potassium cyclopropyltrifluoroborate, potassium cyclopropyltrifluoroboranuide, potassium cyclopropyltrifluororate, potassium cyclopropytrifluoroborate, potassium cycloproyltrifluoroborate, potassiumcyclopropyltrifluoroborate, potassium cyclopropyl-trifluoroborate, cyclopropyltrifluoroborate potassium salt, potassium;cyclopropyl trifluoro boranuide, potassium ion cyclopropyltrifluoroboranuide
23697338
potassium;cyclopropyl(trifluoro)boranuide
[B-](C1CC1)(F)(F)F.[K+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 348°C to 350°C
Color White
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 147.98
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Potassium cyclopropyltrifluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'