missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.05 N (N/20) in Methanol, Ricca Chemical

Product Code. 31940495
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Natural Poly Bottle
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31940495 1 L Natural Poly Bottle 1 L
31940488 4 L Natural Poly Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31940495 Supplier Ricca Chemical Company Supplier No. R62668001A
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1310-58-3 , 67-56-1 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, columbian spirit
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Min % 3.86, 94.0
CAS Max % 4.02, 98.0
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Methanol
pH >12
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.05 Normal
Chemical Name or Material Potassium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.