missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide Solution, 0.1M, Chem-Lab

Catalog Number p-8014402
Click to view available options
Solvent or Matrix:
Ethanol
IPA (2-Propanol)
Water
pH:
13
14
This item is not returnable. View return policy
1310-58-3, 64-17-5
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
CHEBI:32035
[OH-].[K+]
This item is not returnable. View return policy
1310-58-3, 64-17-5
HKO
56.11
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
CHEBI:32035
[OH-].[K+]

Used as a titrant in volumetric analysis. Potassium Hydroxide Solution, 0.1M, Chem-Lab is a laboratory reagent that has 5.612g KOH per 1L of solvent. It is highly acidic in nature and is highly corrosive when handled. Choice of alcohol solvents or water matrix.

Potassium hydroxide solution is an effective conductor and alcoholic solutions are highly flammable. It is used in the production of other potassium compounds and as an electrolyte in alkaline batteries, such as those used in television remote controls. It is used in the manufacture of soft soaps and the production of biodiesel. It can be also be used in disinfection agents for surfaces that resist KOH corrosion, in chemical cremation and to identify some species of fungi.
pH 14
Color Colorless
Boiling Point 79°C
Melting Point -117°C
Concentration 5.612g KOH/1L solvent
Density 0.84g/mL
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Soluble in Water
Quantity 1 L
IUPAC Name potassium hydroxide
Explosion Limit 3.5–15.0% (V)
Flash Point 12°C
Formula Weight 46.07g/mol
Normality 0.1 N (±0.0002/20°C)
Odor Ethanolic
Solvent or Matrix Ethanol
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hydroxide 0.1 mol/l in Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide,ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'