Learn More
Potassium Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™
High quality chemicals for laboratory and specialized industrial use
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Oxidising solid Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use to extinguish.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.