missing translation for 'onlineSavingsMsg'
Learn More

Potassium (tert-butoxymethyl)trifluoroborate, 97%, ACROS Organics™

Brand:  Acros Organics 444840050

Additional Details : CAS : 910251-10-4 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11906081

  • € 374.50 / 5g


Estimated Shipment: 06-07-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

910251-10-4
194.046
potassium tert-butoxymethyl trifluoroborate, potassium tert-butoxymethyltrifluoroborate, potassium tert-butoxymethyl trifluoroboranuide, potassium tert-butoxy methyl trifluoroboranuide, potassium tert-butoxymethyl trifluoro borate 1-
potassium;trifluoro-[(2-methylpropan-2-yl)oxymethyl]boranuide
C5H11BF3KO
XBEPCFNGTDJTIY-UHFFFAOYSA-N
45588181
[B-](COC(C)(C)C)(F)(F)F.[K+]

Specifications

Potassium (tert-butoxymethyl)trifluoroborate
910251-10-4
C5H11BF3KO
XBEPCFNGTDJTIY-UHFFFAOYSA-N
potassium;trifluoro-[(2-methylpropan-2-yl)oxymethyl]boranuide
45588181
5g
97%
potassium tert-butoxymethyl trifluoroborate, potassium tert-butoxymethyltrifluoroborate, potassium tert-butoxymethyl trifluoroboranuide, potassium tert-butoxy methyl trifluoroboranuide, potassium tert-butoxymethyl trifluoro borate 1-
[B-](COC(C)(C)C)(F)(F)F.[K+]
194.046
194.05
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Potassium (tert-butoxymethyl)trifluoroborate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates