missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POTEE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560301
This item is not returnable.
View return policy
Description
Immunogen sequence: LKALQLNELT MDDDTAVLVI DNGS Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 46%.
The function of this protein remains unknown.
Specifications
| POTEE | |
| Polyclonal | |
| Unconjugated | |
| POTEE | |
| A26C1; A26C1A; ANKRD26-like family C member 1A; ANKRD26-like family C, member 1A; cancer/testis antigen family 104, member 2; CT104.2; POTE ankyrin domain family member E; POTE ankyrin domain family, member E; POTE2; POTE-2; POTE2gamma; POTEE; Prostate, ovary, testis-expressed protein on chromosome 2; protein expressed in prostate, ovary, testis, and placenta 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 445582 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6S8J3 | |
| POTEE | |
| Recombinant protein corresponding to Human POTEE. Recombinant protein control fragment (Product #RP-104357). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction