Learn More
POTEG Antibody (OTI2G1), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-73540V
Product Code. 18110287
  • € 751.00 / 0.10mL
Description
POTEG Monoclonal specifically detects POTEG in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POTEG
Monoclonal
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
POTEG
Full length recombinant protein of human POTEG (NP_001005356) produced in HEK293T cells.
0.1 mL
404785.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI2G1
Western Blot, Immunohistochemistry
A26C2, ANKRD26-like family C, member 2, cancer/testis antigen family 104, member 4, CT104.4, POTE ankyrin domain family member G, POTE ankyrin domain family, member G, POTE14ACTBL1, POTE14alpha, POTE-14ANKRD26-like family C member 2, POTE22, Prostate, ovary, testis-expressed protein on chromosome 14, protein expressed in prostate, ovary, testis, and placenta 14
Mouse
Affinity Purified