Learn More
POTEG Monoclonal Antibody (OTI2G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808095S
This item is not returnable.
View return policy
Description
POTEG Monoclonal antibody specifically detects POTEG in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| POTEG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6S5H5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 404785 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G1 | |
| Unconjugated | |
| POTEG | |
| A26C2; ACTBL1; ANKRD26-like family C member 2; ANKRD26-like family C, member 2; cancer/testis antigen family 104, member 4; CT104.4; POTE ankyrin domain family member G; POTE ankyrin domain family, member G; POTE14; POTE-14; POTE14alpha; POTE22; POTEG; Prostate, ovary, testis-expressed protein on chromosome 14; protein expressed in prostate, ovary, testis, and placenta 14 | |
| Full length human recombinant protein of human POTEG produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |