missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU2AF1 Monoclonal Antibody (OTI13G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807999
This item is not returnable.
View return policy
Description
B cell Oct binding protein 1 (BOB-1), is a transcription factor that is required for germinal center formation and immunoglobulin production. BOB-1 is expressed at high levels in spleen and peripheral blood leukocytes and represents an OCT co-factor capable of conferring cell specific activation of OCT-1 and OCT-2.Specifications
| POU2AF1 | |
| Monoclonal | |
| Unconjugated | |
| POU2AF1 | |
| BOB1, OBF-1, OBF1, OCAB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5450 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI13G1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q16633 | |
| POU2AF1 | |
| Full length human recombit protein of human POU2AF1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction