Learn More
POU2AF1 Monoclonal Antibody (OTI13G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807999S
This item is not returnable.
View return policy
Description
POU2AF1 Monoclonal antibody specifically detects POU2AF1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| POU2AF1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O96009 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9476 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI13G1 | |
| Unconjugated | |
| NAPSA | |
| B-cell-specific coactivator OBF-1; BOB.1; Bob1; BOB-1; OBF.1; OBF1; Obf-1; OCAB; OCA-B; OCT-binding factor 1; POU class 2 associating factor 1; POU domain class 2-associating factor 1; POU domain, class 2, associating factor 1; Pou2af1 | |
| Human recombinant protein fragment corresponding to amino acids 64-244 of human NAPSA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |