Learn More
POU2AF1 Mouse anti-Human, Clone: OTI1E11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF806396
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
B cell Oct binding protein 1 (BOB-1), is a transcription factor that is required for germinal center formation and immunoglobulin production. BOB-1 is expressed at high levels in spleen and peripheral blood leukocytes and represents an OCT co-factor capable of conferring cell specific activation of OCT-1 and OCT-2.Specifications
| POU2AF1 | |
| Monoclonal | |
| Unconjugated | |
| POU2AF1 | |
| BOB1, OBF-1, OBF1, OCAB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5450 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1E11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q16633 | |
| POU2AF1 | |
| Full length human recombit protein of human POU2AF1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.