Learn More
POU3F2/OCT7 Antibody (CL6232) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61437JF669
Product Code. 30302127
  • € 737.00 / 0.10mL
Description
POU3F2/OCT7 Monoclonal antibody specifically detects POU3F2/OCT7 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POU3F2/OCT7
Monoclonal
Janelia Fluor 669
Brain-2, Brain-specific homeobox/POU domain protein 2, brn-2, BRN2brain-2, Nervous system-specific octamer-binding transcription factor N-Oct-3, Oct-7, OCT7POU domain class 3, transcription factor 2, Octamer-binding protein 7, Octamer-binding transcription factor 7, OTF7, OTF-7, POU class 3 homeobox 2, POU domain, class 3, transcription factor 2, POUF3
This antibody was developed using a recombinant protein derived from P20265, with the exact immunogen sequence remaining proprietary.
0.1 mL
Transcription Factors and Regulators
5454
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL6232
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat