missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POU3F3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570366
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 92%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
POU3F3 is a member of the class III POU family of transcription factors that are expressed in the central nervous system. The POU domain in these proteins is required for high affinity binding to octamer DNA sequences.
Specifications
| POU3F3 | |
| Polyclonal | |
| Unconjugated | |
| POU3F3 | |
| 8-Oct; Brain-1; brain-specific homeobox/POU domain protein 1; Brn1; Brn-1; brn-1 protein; class III POU protein POU-domain; oct-8; Octamer-binding protein 8; octamer-binding transcription factor 8; Otf8; OTF-8; POU class 3 homeobox 3; POU domain, class 3, transcription factor 3; Pou3f3; RHS1; Rhs2; Skin1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 192109, 5455 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P20264, Q63262 | |
| POU3F3 | |
| synthetic peptide directed towards the C-terminal region of rat POU3F3. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction