missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU4F3 Antibody (OTI4C5), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73541AF350
This item is not returnable.
View return policy
Description
POU4F3 Monoclonal specifically detects POU4F3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| POU4F3 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| POU4F3 | |
| Recombinant protein fragment corresponding to amino acids 179-338 of human POU4F3 (NP_002691) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI4C5 | |
| Western Blot | |
| brain-3C, Brain-specific homeobox/POU domain protein 3C, BRN3.1, Brn-3C, BRN3Cbrn-3C, DFNA15, MGC138412, POU class 4 homeobox 3, POU domain class 4, transcription factor 3, POU domain, class 4, transcription factor 3 | |
| Mouse | |
| Affinity Purified | |
| Neuroscience, Vision | |
| 5459.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction