Learn More
POU4F3 Monoclonal Antibody (OTI3A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808731
Encompass_Preferred
 View more versions of this product
Product Code. 17560294
  • € 540.87 / 100µL
Description
This gene encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal domit type 15.
Specifications
Specifications
POU4F3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15319
POU4F3
Human recombit protein fragment corresponding to amino acids 179-338 of human POU4F3 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI3A12
Unconjugated
POU4F3
BRN3C, DFNA15, DFNA42, DFNA52
Mouse
Affinity Chromatography
RUO
5459
-20° C, Avoid Freeze/Thaw Cycles