missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU4F3 Monoclonal Antibody (OTI4C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808720
This item is not returnable.
View return policy
Description
This gene encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal domit type 15.Specifications
| POU4F3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15319 | |
| POU4F3 | |
| Human recombit protein fragment corresponding to amino acids 179-338 of human POU4F3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4C5 | |
| Unconjugated | |
| POU4F3 | |
| BRN3C, DFNA15, DFNA42, DFNA52 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5459 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction