missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU4F3 Monoclonal Antibody (OTI4C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808720S
This item is not returnable.
View return policy
Description
POU4F3 Monoclonal antibody specifically detects POU4F3 in Human samples. It is validated for Western BlotSpecifications
| POU4F3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q53T94 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9014 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4C5 | |
| Unconjugated | |
| TAF1B | |
| brain POU domain gene 3.1; brain-3.1; Brain-3C; brain-specific homeobox POU domain protein 3C; brain-specific homeobox/POU domain protein 3.1; Brain-specific homeobox/POU domain protein 3C; brn3.1; Brn-3.1; brn3c; brn-3c; ddl; DFNA15; dreidel; MGC138412; POU class 4 homeobox 3; POU domain transcription factor Brn3C; POU domain, class 4, transcription factor 3; pou4f3; zfbrn-3.1 | |
| Human recombinant protein fragment corresponding to amino acids 1-208 of human TAF1B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction