missing translation for 'onlineSavingsMsg'
Learn More

POU4F3 Monoclonal Antibody (OTI6H9), TrueMAB™, OriGene

Product Code. 17530344
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17530344 30 μL 30µL
17510344 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17530344 Supplier Origene Technologies Supplier No. TA808755S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POU4F3 Monoclonal antibody specifically detects POU4F3 in Human samples. It is validated for Western Blot

Specifications

Antigen POU4F3
Applications Western Blot
Classification Monoclonal
Clone OTI6H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MAGEB3
Gene Accession No. O15480
Gene Alias brain POU domain gene 3.1; brain-3.1; Brain-3C; brain-specific homeobox POU domain protein 3C; brain-specific homeobox/POU domain protein 3.1; Brain-specific homeobox/POU domain protein 3C; brn3.1; Brn-3.1; brn3c; brn-3c; ddl; DFNA15; dreidel; MGC138412; POU class 4 homeobox 3; POU domain transcription factor Brn3C; POU domain, class 4, transcription factor 3; pou4f3; zfbrn-3.1
Host Species Mouse
Immunogen Full length human recombinant protein of human MAGEB3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4114
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.