Learn More
Invitrogen™ POU4F3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA540644
Product Code. 15554302
  • € 516.00 / 100µL
Description
Peptide sequence: ALANLKIPGV GSLSQSTICR FESLTLSHNN MIALKPVLQA WLEEAEAAYR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
POU4F3 encodes a member of the POU-domain family of transcription factors. POU-domain proteins have been observed to play important roles in control of cell identity in several systems. This protein is found in the retina and may play a role in determining or maintaining the identities of a small subset of visual system neurons. Defects in this gene are the cause of non-syndromic sensorineural deafness autosomal dominant type 15.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
POU4F3
Polyclonal
Unconjugated
Pou4f3
brain POU domain gene 3.1; brain-3.1; Brain-3C; brain-specific homeobox POU domain protein 3C; brain-specific homeobox/POU domain protein 3.1; Brain-specific homeobox/POU domain protein 3C; brn3.1; Brn-3.1; brn3c; brn-3c; ddl; DFNA15; dreidel; MGC138412; POU class 4 homeobox 3; POU domain transcription factor Brn3C; POU domain, class 4, transcription factor 3; pou4f3; zfbrn-3.1
Rabbit
Protein A
RUO
18998, 5459
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q15319, Q63955
Pou4f3
Synthetic peptide directed towards the middle region of human POU4F3.
100 μL
Primary
Human, Mouse