missing translation for 'onlineSavingsMsg'
Learn More
Learn More
POU5F2, Mouse anti-Human, Clone: 1B12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00134187-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MPLRVDTLTWLSTQAAPGRVMVWPAVRPGICPGPDVWRIPLGPLPHEFRGWIAPCRPRLGASEAGDWLRRPSEGALPGPYIALRSIPKLPPPEDISGILKELQQLAKELRQKRLSLGYSQADVGIAVGALFGKVLSQTTICRFEAQQLSVANMWKLRPLLKKWLKEVEAENLLGLCKMEMILQQSGKWRRASRERRIGNSLEKFFQRCPKPTPQQISHIAGCLQLQKDVVRVWFYNRSKMGSRPTNDASPREIVGTAGPPCPGAPVCFHLGLGLPVDIPHYTRLYSAGVAHSSAPATTLGLLRFSpecifications
| POU5F2 | |
| Monoclonal | |
| Unconjugated | |
| POU domain class 5, transcription factor 2 | |
| DKFZp686P02123/FLJ25680/SPRM-1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1B12 | |
| In 1x PBS, pH 7.4 | |
| BC029532 | |
| POU5F2 | |
| POU5F2 (AAH29532, 1 a.a. ∼ 304 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 134187 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction