Learn More
Invitrogen™ POU5F2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139955
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: QQLSVANMWK LRPLLKKWLK EVEAENLLGL CKMEMILQQS GKWRRASRER RIGNSLEKFF QRCPKP Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 73%.
Transcription factor that binds preferentially to the octamer motif (5'-ATGTTAAT-3'). May exert a regulatory function in meiotic events that are required for terminal differentiation of male germ cell.
Specifications
| POU5F2 | |
| Polyclonal | |
| Unconjugated | |
| POU5F2 | |
| 1700013G10Rik; POU domain class 5, transcription factor 2; POU domain, class 5, transcription factor 2; Pou5f2; Sperm 1 POU domain transcription factor; sperm-associated POU domain protein 1; Sprm1; SPRM-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 134187 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N7G0 | |
| POU5F2 | |
| Recombinant protein corresponding to Human POU5F2. Recombinant protein control fragment (Product #RP-109638). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.