missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POU6F1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537884
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat, dog and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Brn-5 (Brain-5), also known as POU6F1, MPOU or TCFB1, is a widely expressed protein but during embryogenesis is exclusively found in the developing brain and spinal cord. As is characteristic of Brn family members, Brn-5 contains two DNA-binding domains, namely the POU-specific domain and the POU homeodomain, which each contain an HTH (helix-turn-helix) motif. Brn-5 binds to CRH (corticotrophin-releasing hormone) elements with high affinity and is capable of both enhancing Prolactin gene expression and activating Pit-1 expression.
Specifications
| POU6F1 | |
| Polyclonal | |
| Unconjugated | |
| POU6F1 | |
| 2310038G18Rik; brain-5; Brain-specific homeobox/POU domain protein 5; BRN5; brn-5; brn-5 protein; C79593; cns-1; Emb; MPOU; mPOU homeobox protein; Octamer-binding transcription factor EMB; POU class 6 homeobox 1; POU domain, class 6, transcription factor 1; Pou6f1; TCFB1; transcription regulatory protein MCP-1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 19009, 5463 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q07916, Q14863 | |
| POU6F1 | |
| Peptide with sequence C-ETPTVSQLVSKPHTP, from the internal region of POU6F1 (aa 121-135). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction