missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POU6F1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565617

Product Code. 15908364

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: KPSASAPIPIT CSETPTVSQL VSKPHTPSLD EDGINLEEI Highest antigen sequence identity to the following orthologs - mouse 95%, rat 100%.

Brn-5 (Brain-5), also known as POU6F1, MPOU or TCFB1, is a widely expressed protein but during embryogenesis is exclusively found in the developing brain and spinal cord. As is characteristic of Brn family members, Brn-5 contains two DNA-binding domains, namely the POU-specific domain and the POU homeodomain, which each contain an HTH (helix-turn-helix) motif. Brn-5 binds to CRH (corticotrophin-releasing hormone) elements with high affinity and is capable of both enhancing Prolactin gene expression and activating Pit-1 expression.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

POU6F1
Polyclonal
Unconjugated
POU6F1
2310038G18Rik; brain-5; Brain-specific homeobox/POU domain protein 5; BRN5; brn-5; brn-5 protein; C79593; cns-1; Emb; MPOU; mPOU homeobox protein; Octamer-binding transcription factor EMB; POU class 6 homeobox 1; POU domain, class 6, transcription factor 1; Pou6f1; TCFB1; transcription regulatory protein MCP-1
Rabbit
Antigen affinity chromatography
RUO
5463
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q14863
POU6F1
Recombinant Human POU6F1. Recombinant protein control fragment (Product #RP-106284).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.