missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ POU6F1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15943705
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15943705 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15943705 Supplier Invitrogen™ Supplier No. PA577195

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Brn-5 (Brain-5), also known as POU6F1, MPOU or TCFB1, is a widely expressed protein but during embryogenesis is exclusively found in the developing brain and spinal cord. As is characteristic of Brn family members, Brn-5 contains two DNA-binding domains, namely the POU-specific domain and the POU homeodomain, which each contain an HTH (helix-turn-helix) motif. Brn-5 binds to CRH (corticotrophin-releasing hormone) elements with high affinity and is capable of both enhancing Prolactin gene expression and activating Pit-1 expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POU6F1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene POU6F1
Gene Accession No. P56223, Q07916, Q14863
Gene Alias 2310038G18Rik; brain-5; Brain-specific homeobox/POU domain protein 5; BRN5; brn-5; brn-5 protein; C79593; cns-1; Emb; MPOU; mPOU homeobox protein; Octamer-binding transcription factor EMB; POU class 6 homeobox 1; POU domain, class 6, transcription factor 1; Pou6f1; TCFB1; transcription regulatory protein MCP-1
Gene Symbols POU6F1
Host Species Rabbit
Immunogen Recombinant full length Human POU6F1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116545, 19009, 5463
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.