missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pou6f2 Polyclonal Antibody

Beskrivning
This antibody is predicted to react with bovine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a member of the POU protein family characterized by the presence of a bipartite DNA binding domain, consisting of a POU-specific domain and a homeodomain, separated by a variable polylinker. The DNA binding domain may bind to DNA as monomers or as homo- and/or heterodimers, in a sequence-specific manner. The POU family members are transcriptional regulators, many of which are known to control cell type-specific differentiation pathways. This gene is a tumor suppressor involved in Wilms tumor (WT) predisposition. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifikationer
Specifikationer
| Antigen | Pou6f2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Pou6f2 |
| Gene Accession No. | Q8BJI4 |
| Gene Alias | D130006K24Rik; POU class 6 homeobox 2; POU domain class 6 transcription factor 2; POU domain, class 6, transcription factor 2; Pou6f2; Retina-derived POU domain factor 1; retina-derived POU-domain factor-1; RPF1; RPF-1; Wilms tumor suppressor locus; WT5; WTSL |
| Gene Symbols | Pou6f2 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?