Learn More
Labconco™ Power Cord 10A/230V
Shop All Labconco Corporation Products
Click to view available options
For Use With (Equipment):
Logic Equipment, Freeze Dryers
Unit Size:
Each
Description
- Power Cord with molded plug and connector for Continental Europe (Schuko)
- 3 m length
Specifications
Specifications
| Type | Power Cord |
| For Use With (Equipment) | Logic Equipment, Freeze Dryers |
| Length (Metric) | 3 m |
| Voltage | 230 V |
| Amperage | 10 A |