Learn More
Uvex™ Power Protect V1000 Safety Glove
Catalog Number 17797629
Click to view available options
Size:
7
8
9
10
11
The Uvex power protect V1000 protective glove offers safe protection against electrical voltages up to 1000 V.
  • The glove's high level of flexibility offers pleasant wearer comfort and an excellent tactile feel.
  • Anatomical fit
  • Flexible, with a good grip feel even at low temperatures
  • Flexible at cold temperatures
  • Mineral oil-resistant
Certifications/Compliance EN 60903: 2003 IEC 60903: 2014 (Class 0/RC)
Color Red
Material Latex
Size 11