Learn More
Uvex™ Power Protect V1000 Safety Glove
Click to view available options
Size:
7
8
9
10
11
Description
- The glove's high level of flexibility offers pleasant wearer comfort and an excellent tactile feel.
- Anatomical fit
- Flexible, with a good grip feel even at low temperatures
- Flexible at cold temperatures
- Mineral oil-resistant
Specifications
Certifications/Compliance | EN 60903: 2003 IEC 60903: 2014 (Class 0/RC) |
Color | Red |
Material | Latex |
Size | 11 |