Learn More
Thermo Scientific Power Supply Cable for Lynx Superspeed
IEC60309 32A-6h-3-pin Blue, 200/250 V
Brand: Thermo Scientific 20190357
Additional Details : Weight : 1.00000kg
Description
- IEC60309 32A-6h-3-pin Blue, 200/250 V
Specifications
Power Cable | |
Reusable | |
IEC60309 32A-6h-3-pin Blue, 200/250 V |
Centrifuge | |
Blue | |
200/250 V |