Learn More
3M™ Powerflow™ Full Facepiece 7900
Click to view available options
:
Color: Black
Description
- Replacement full face mask for 3M™ Powerflow™ 7900PF Powered Air Respirator
- Speech diaphragm to improve communication
- For use with 3M™ Powered Air Respirator 7900PF
Specifications
Specifications
| Certifications/Compliance | EN 136:1998 (Class 2), CE Approved |
| Color | Black |
| Material | Silicone, Polycarbonate |
| For Use With (Equipment) | 3M™ Powerflow™ 7900PF Powered Air Respirator |
| Mounting Type | Face-mounted |