Learn More
Poulten & Graf GmbH PP-Stopper
Poulten & Graf GmbH PP-Stopper uses a sealing profile to secure the neck of a flask or bottle.
Brand:  Poulten & Graf GmbH 10144415
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10137961
  • € 0.90 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Polypropylene
  • With sealing profile
  • According to DIN 12254
Specifications
Specifications
PP-Stopper
10
Flasks and bottles
DIN 12254
TS 12/21
Polypropylene
TS 12/21