Learn More
Poulten & Graf GmbH PP-Stopper
Poulten & Graf GmbH PP-Stopper uses a sealing profile to secure the neck of a flask or bottle.
Brand: Poulten & Graf GmbH 10144415
Additional Details : Weight : 0.10000kg
Description
- Polypropylene
- With sealing profile
- According to DIN 12254
Specifications
PP-Stopper | |
10 | |
Flasks and bottles | |
DIN 12254 |
TS 12/21 | |
Polypropylene | |
TS 12/21 |