Learn More
Poulten & Graf GmbH PP-Stopper
Click to view available options
Size:
TS 10/19
TS 12/21
TS 24/29
TS 29/32
TS14/23
Unit Size:
Each
Description
- Polypropylene
- With sealing profile
- According to DIN 12254
Specifications
Specifications
| Certifications/Compliance | DIN 12254 |
| Description | PP-Stopper |
| For Use With (Equipment) | Flasks and bottles |
| Quantity | 10 |
| Size | TS 10/19 |
| Standard Taper Joint | TS 10/19 |
| Material | Polypropylene |