missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP1 alpha/PPP1A Antibody (4G3), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04334JF646
This item is not returnable.
View return policy
Description
PP1 alpha/PPP1A Monoclonal antibody specifically detects PP1 alpha/PPP1A in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PP1 alpha/PPP1A | |
| Monoclonal | |
| Janelia Fluor 646 | |
| EC 3.1.3.16, PP1alpha, PP-1AMGC15877, PPP1AMGC1674, protein phosphatase 1, catalytic subunit, alpha isoform, protein phosphatase 1, catalytic subunit, alpha isozyme, serine/threonine protein phosphatase PP1-alpha 1 catalytic subunit, serine/threonine-protein phosphatase PP1-alpha catalytic subunit | |
| Recombinant human PP1 alpha/PPP1A (30-299aa) purified from E. coli | |
| 0.1 mL | |
| Protein Phosphatase | |
| 5499 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunofluorescence | |
| 4G3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction