missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP1 gamma Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112122
This item is not returnable.
View return policy
Description
This antibody reacts with: Human, Mouse, Rat, Bovine, Xenopus tropicalis (Species predicted to react based on 100% sequence homology).
The protein encoded by this gene belongs to the protein phosphatase family, PP1 subfamily. PP1 is an ubiquitous serine/threonine phosphatase that regulates many cellular processes, including cell division. It is expressed in mammalian cells as three closely related isoforms, alpha, beta/delta and gamma, which have distinct localization patterns. This gene encodes the gamma isozyme. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| PP1 gamma | |
| Polyclonal | |
| Unconjugated | |
| PPP1CC | |
| dis2m1; MGC1675; MGC2870; PP1; PP1C; PP1C gamma 1; PP1C gamma 2; PP1G; PP-1G; PP1gamma; PP2CG; PP2CGAMMA; PPP1CC; Ppp1cc1; PPP1G; PPP2CG; protein phosphatase 1 catalytic subunit gamma; protein phosphatase 1, catalytic subunit, gamma isoform; protein phosphatase 1, catalytic subunit, gamma isozyme; protein phosphatase 1C catalytic subunit; protein serine-threonine phosphatase catalytic subunit PP-1b; serine/threonine phosphatase 1 gamma; serine/threonine-protein phosphatase PP1-gamma catalytic subunit | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5501 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with no preservative | |
| P36873 | |
| PPP1CC | |
| A synthetic peptide corresponding to the center region of the Human PPP1CC. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction