Learn More
PP14/Glycodelin Mouse anti-Human, Clone: 16, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-05115-0.1mg
 View more versions of this product
Product Code. 18582611
  • € 419.00 / 0.10mg
Description
PP14/Glycodelin Monoclonal specifically detects PP14/Glycodelin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PP14/Glycodelin
Monoclonal
1 mg/mL
ELISA 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin
alpha uterine protein, GdF, GdS, glycodelin, glycodelin-A, glycodelin-F, glycodelin-S, MGC138509, MGC142288, PEP, Placental protein 14, PP14 protein (placental protein 14), PP14GDPAEGGdA, pregnancy-associated endometrial a, Pregnancy-associated endometrial alpha-2 globulin, pregnancy-associated endometrial alpha-2-globulin, progestagen-associated endometrial protein (placental protein 14, progestagen-associated endometrial proteinPEG, Progesterone-associated endometrial protein
Placental protein 14 (glycodelin A) purified from second trimester amniotic fluid
0.1 mg
Primary
Human
Purified
ELISA
16
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl
Mouse
Protein A or G purified
RUO
5047
Store at 4°C. Do not freeze.
IgG1 κ