missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP14B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589977
This item is not returnable.
View return policy
Description
Immunogen sequence: RLNLEEWILE QLTRLYDCQE EEIPELEIDV DELLDMESDD ARAARVKELL VDCYKPTEAF ISGLLDKIRG MQKLSTPQKK; Positive Samples: HeLa, A-431; Cellular Location: Cytoplasm.
This gene is thought to play a role in a complex developmental pathway and the regulation of genes involved in cerebellar development. Mutations in this gene have been associated with autosomal recessive spinocerebellar ataxia.
Specifications
| PP14B | |
| Polyclonal | |
| Unconjugated | |
| PPP1R14B | |
| AOM172; Phi1; PHI-1; Phosphatase holoenzyme inhibitor 1; phospholipase C beta-3 neighboring gene protein; phospholipase c neighboring; phospholipase C-beta-3 neighbouring gene protein; PLCB3N; PNG; Ppp1r14b; protein phosphatase 1 regulatory inhibitor subunit 14B; protein phosphatase 1 regulatory subunit 14B; protein phosphatase 1, regulatory (inhibitor) 5 subunit 14B; protein phosphatase 1, regulatory (inhibitor) subunit 14B; SOM172; ubiquitous PKC-potentiated PP1 inhibitor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18938, 259225, 26472 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.71 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q62084, Q8K3F3, Q96C90 | |
| PPP1R14B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 68-147 of human PPP1R14B (NP_6196341). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction