missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2A alpha Antibody (H5D12), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29818AF488
This item is not returnable.
View return policy
Description
PP2A alpha Monoclonal antibody specifically detects PP2A alpha in Human samples. It is validated for Western Blot
Specifications
| PP2A alpha | |
| Monoclonal | |
| Alexa Fluor 488 | |
| EC 3.1.3.16, PP2A-alpha, PP2Ac, PP2CA, PP2Calpha, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isozyme, protein phosphatase 2A catalytic subunit, alpha isoform, Replication protein C, RP-C, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform, serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform | |
| GST fusion protein corresponding to the N-terminus of human PP2A, B subunit, B56 delta. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Neuronal Cell Markers, Neurotransmission, Protein Phosphatase, Signal Transduction | |
| 5515 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| H5D12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction