missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PP2A alpha/beta Recombinant Rabbit Monoclonal Antibody (4J2Z8)
GREENER_CHOICE

Product Code. 17267033 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17267033 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17267033 Supplier Invitrogen™ Supplier No. MA538160

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, 293T, NIH/3T3, Jurkat, C6, Mouse testis, Mouse brain, Rat lung, Rat testis Immunogen sequence: WSDPDDRGGW GISPRGAGYT FGQDISETFN HANGLTLVSR AHQLVMEGYN WCHDRNVVTI FSAPNYCYRC GNQAAIMELD DTLKYSFLQF DPAPRRGEPH VTRRTPDYFL.

This gene encodes the phosphatase 2A catalytic subunit. Protein phosphatase 2A is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. This gene encodes an alpha isoform of the catalytic subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PP2A alpha/beta
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4J2Z8
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPP2CB
Gene Accession No. P62714, P62715, P62716, P63330, P63331, P67775
Gene Alias AI115466; D8Ertd766e; fb17h07; fc05c02; fk24a01; hypothetical protein LOC550585; PP2A; Pp2a1; Pp2a2; PP2A-alpha; PP2Abeta; PP2A-beta; PP2Ac; PP2CA; PP2Calpha; PP2CB; ppp2ca; ppp2caa; PPP2CB; protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform; protein phosphatase 2 (formerly 2A), catalytic subunit, beta isoform; protein phosphatase 2 catalytic subunit alpha; protein phosphatase 2 catalytic subunit alpha a; protein phosphatase 2 catalytic subunit beta; protein phosphatase 2, catalytic subunit, alpha isoform; protein phosphatase 2, catalytic subunit, alpha isozyme; protein phosphatase 2, catalytic subunit, beta isoform; protein phosphatase 2, catalytic subunit, beta isozyme; protein phosphatase 2,catalytic subunit alpha isozyme; protein phosphatase 2A alpha subunit; protein phosphatase 2A beta subunit; protein phosphatase 2A catalytic alpha subunit; protein phosphatase 2A catalytic subunit alpha isoform; protein phosphatase 2A catalytic subunit beta isoform; protein phosphatase 2A catalytic subunit, beta isoform; protein phosphatase 2a, catalytic subunit, alpha isoform; protein phosphatase 2A, catalytic subunit, beta isoform; protein phosphatase type 2A catalytic subunit; protein phosphatase-2A-alpha; protein phosphatase-2A-beta; R75353; replication protein C; RP C; RP-C; serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform; serine/threonine protein phosphatase 2A, catalytic subunit, beta isoform; serine/threonine-protein phosphatase 2A catalytic subunit alpha isoform; Serine/threonine-protein phosphatase 2A catalytic subunit beta isoform; testicular tissue protein Li 146; wu:fb17h07; wu:fc05c02; wu:fk24a01; zgc:110641; zgc:55808; zgc:77172
Gene Symbols PPP2CA, PPP2CB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-309 of human PP2A alpha + beta (P62714).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19052, 19053, 24672, 24673, 5515, 5516
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.