missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2C alpha/PPM1A Antibody (7F12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | PP2C alpha/PPM1A |
|---|---|
| Clone | 7F12 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645985
|
Novus Biologicals
NBP2-37480-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18164408
|
Novus Biologicals
NBP2-37480 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP2C alpha/PPM1A Monoclonal specifically detects PP2C alpha/PPM1A in Human, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| PP2C alpha/PPM1A | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human, Primate, Monkey | |
| EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
| PPM1A | |
| IgG1 | |
| Affinity Purified |
| 7F12 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| 5494 | |
| Purified recombinant fragment of human PPM1A (AA: 202-382) expressed in E. Coli. | |
| Primary | |
| 42.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title