missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2C alpha/PPM1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | PP2C alpha/PPM1A |
|---|---|
| Dilution | Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663586
|
Novus Biologicals
NBP3-00058-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625165
|
Novus Biologicals
NBP3-00058-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PP2C alpha/PPM1A Polyclonal antibody specifically detects PP2C alpha/PPM1A in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| PP2C alpha/PPM1A | |
| Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 5494 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Immunoprecipitation 1-5uL/mg of lysate, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.1.3.16, FLJ42306, MGC9201, phosphatase 2C alpha, PP2CA, PP2Calpha, PP2C-ALPHA, PPPM1A, protein phosphatase 1A, protein phosphatase 1A (formerly 2C), magnesium-dependent, alpha isoform, protein phosphatase 2C alpha isoform, Protein phosphatase 2C isoform alpha, Protein phosphatase IA, protein phosphatase, Mg2+/Mn2+ dependent, 1A | |
| Produced in rabbits immunized with purified, recombinant Human PP2C alpha/PPM1A (Accession#: P35813-1; Met1-Trp382) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title