missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2C beta/PPM1B Antibody (OTI3F2), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73547AF405
This item is not returnable.
View return policy
Description
PP2C beta/PPM1B Monoclonal specifically detects PP2C beta/PPM1B in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| PP2C beta/PPM1B | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PPM1B | |
| Full length recombinant protein of human PP2C beta/PPM1B (NP_002697) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry | |
| OTI3F2 | |
| Western Blot, Flow Cytometry | |
| EC 3.1.3.16, MGC21657, PP2C-beta, PP2C-beta-X, PP2CBmagnesium-dependent, beta isoform, protein phosphatase 1B, Protein phosphatase 2C isoform beta, protein phosphatase 2C-like protein, protein phosphatase, Mg2+/Mn2+ dependent, 1B | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 5495.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction