missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP2C gamma/PPM1G Antibody (k1G6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04337CL1
This item is not returnable.
View return policy
Description
PP2C gamma/PPM1G Monoclonal antibody specifically detects PP2C gamma/PPM1G in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| PP2C gamma/PPM1G | |
| Monoclonal | |
| CoraFluor 1 | |
| EC 3.1.3.16, MGC1675, MGC2870, PP2C, gamma, PP2CG, PP2C-gamma, PP2CGAMMA, PPM1C, PPP2CG, Protein phosphatase 1C, protein phosphatase 1G, protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, protein phosphatase 2, catalytic subunit, gamma isoform, protein phosphatase 2C gamma isoform, Protein phosphatase 2C isoform gamma, Protein phosphatase magnesium-dependent 1 gamma, protein phosphatase, Mg2+/Mn2+ dependent, 1G | |
| Recombinant human PP2C gamma/PPM1G (317-546aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 5496 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| k1G6 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction