missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PP4/PPP4C Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99380B
This item is not returnable.
View return policy
Description
PP4/PPP4C Polyclonal antibody specifically detects PP4/PPP4C in Human samples. It is validated for Western Blot
Specifications
| PP4/PPP4C | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| EC 3.1.3.16, PP4C, PP4protein phosphatase X, catalytic subunit, PP-X, PPXPPH3, protein phosphatase 4 (formerly X), catalytic subunit, protein phosphatase 4, catalytic subunit, Protein phosphatase X | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PP4/PPP4C. | |
| 0.1 mL | |
| Core ESC Like Genes, Protein Phosphatase, Signal Transduction, Stem Cell Markers | |
| 5531 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction