missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PP5 Monoclonal Antibody (12F7), APC
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545364
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45364 was sufficient for detection of PP5 in 15 μg of human A431 lysates by ECL analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 58kDa.
This gene encodes a serine/threonine phosphatase which is a member of the protein phosphatase catalytic subunit family. Proteins in this family participate in pathways regulated by reversible phosphorylation at serine and threonine residues; many of these pathways are involved in the regulation of cell growth and differentiation. The product of this gene has been shown to participate in signaling pathways in response to hormones or cellular stress, and elevated levels of this protein may be associated with breast cancer development. Alternative splicing results in multiple transcript variants.
Specifications
| PP5 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| P53041 | |
| Ppp5c | |
| Full length human PP5 protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 12F7 | |
| APC | |
| Ppp5c | |
| ANP receptor; AU020526; EGK_10788; FLJ36922; FLJ55954; OTTHUMP00000165834; PP5; PPP5; Ppp5c; PPT; PP-T; protein phosphatase 5 catalytic subunit; protein phosphatase 5, catalytic subunit; protein phosphatase T; Serine/threonine-protein phosphatase 5 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5536 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction