missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPA1, Mouse anti-Human, Clone: 3B2, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005464-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: AAPFSLEYRVFLKNEKGQYISPFHDIPIYADKDVFHMVVEVPRWSNAKMEIATKDPLNPIKQDVKKGKLRYVANLFPYKGYIWNYGAIPQTWEDPGHNDKHTSpecifications
| PPA1 | |
| Monoclonal | |
| Unconjugated | |
| pyrophosphatase (inorganic) 1 | |
| IOPPP/MGC111556/PP/PP1/SID6-8061 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 3B2 | |
| In 1x PBS, pH 7.4 | |
| NM_021129 | |
| PPA1 | |
| PPA1 (NP_066952, 10 a.a. ∼ 111 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5464 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction