missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPAN Polyclonal Antibody
GREENER_CHOICE

Product Code. 15708141 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15708141 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15708141 Supplier Invitrogen™ Supplier No. PA560302

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RWEMDRGRGR LCDQKFPKTK DKSQGAQARR GPRGASRDGG Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 38%.

The protein encoded by this gene is an evolutionarily conserved protein similar to yeast SSF1 as well as to the gene product of the Drosophila gene peter pan (ppan). SSF1 is known to be involved in the second step of mRNA splicing. Both SSF1 and ppan are essential for cell growth and proliferation. Exogenous expression of this gene was reported to reduce the anchorage-independent growth of some tumor cells. Read-through transcription of this gene with P2RY11/P2Y(11), an adjacent downstream gene that encodes an ATP receptor, has been found. These read-through transcripts are ubiquitously present and up-regulated during granulocyte differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPAN
Gene Accession No. Q9NQ55
Gene Alias A230087P06Rik; Brix domain-containing protein 3; BXDC3; homolog of S. cerevisiae SSF1; Peter Pan homolog; peter pan homolog (Drosophila); Ppan; second-step splicing factor 1; SSF; Ssf1; ssf-1; SSF2; suppressor of sterile four 1; suppressor of SWI4 1 homolog
Gene Symbols PPAN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPAN. Recombinant protein control fragment (Product #RP-99100).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56342
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.