missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAPDC1B/DPPL1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76244F
This item is not returnable.
View return policy
Description
PPAPDC1B/DPPL1 Polyclonal antibody specifically detects PPAPDC1B/DPPL1 in Human, Mouse samples. It is validated for ELISA
Specifications
| PPAPDC1B/DPPL1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA | |
| FITC | |
| diacylglycerol pyrophosphate like 1, DPPL1, EC 3.1.3.4, HTPAPphosphatidate phosphatase PPAPDC1B, phosphatidic acid phosphatase type 2 domain containing 1B, Phosphatidic acid phosphatase type 2 domain-containing protein 1B | |
| Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human PPAPDC1B. The immunogen is located within amino acids 30 - 80 of PPAPDC1B. Amino Acid Squence: LYRNPYVEAEYFPT | |
| 0.1 mL | |
| Signal Transduction | |
| 84513 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction