missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAPDC3 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76246FR
This item is not returnable.
View return policy
Description
PPAPDC3 Polyclonal antibody specifically detects PPAPDC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PPAPDC3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 680 | |
| C9orf67, chromosome 9 open reading frame 67, EC 3.1.3, EC 3.1.3.-, FLJ14662, KIAA0515, MGC12921, NET39, phosphatidic acid phosphatase type 2 domain containing 3, Phosphatidic acid phosphatase type 2 domain-containing protein 3, probable lipid phosphate phosphatase PPAPDC3 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the amino of terminus human PPAPDC3. The immunogen is located within the first 50 amino acids of PPAPDC3. Amino Acid Squence: PPKGGPEPRSSGRKASGPS | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Stem Cells | |
| 84814 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction