missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPAPDC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567093
This item is not returnable.
View return policy
Description
Immunogen sequence: GPYETSPSLLD YLTMDIYAFP AGHASRAAMV SK Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
PPAPDC3, also known as nuclear envelope transmembrane protein 39 (NET39), was initially discovered in an in silico screen for secreted or membrane proteins. It is a member of the PAP2 superfamily of phosphatases and haloperoxidases. PPAPDC3 has recently been shown to act as a negative regulator of myoblast differentiation by diminishing the activity of the mammalian target of rapamycin TOR. PPAPDC3 is highly expressed in cardiac and skeletal muscle and becomes strongly upregulated during cultured myoblast differentiation tissues. Overexpression of PPAPDC3 in myoblasts repressed myogenesis while knockdown by RNA interference promoted differentiation indicating its part in the regulatory mechanism for myogenesis.
Specifications
| PPAPDC3 | |
| Polyclonal | |
| Unconjugated | |
| Plpp7 | |
| C9orf67; D830019K17Rik; Inactive phospholipid phosphatase 7; NET39; Nuclear envelope transmembrane protein 39; nuclear envelope transmembrane protein NET39; phosphatidic acid phosphatase type 2 domain containing 3; phosphatidic acid phosphatase type 2 domain-containing protein 3; phospholipid phosphatase 7 (inactive); PLPP7; Ppapdc3; probable lipid phosphate phosphatase PPAPDC3; RGD1305821; RP11-643E14.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84814 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NBV4 | |
| Plpp7 | |
| Recombinant Human PPAPDC3. Recombinant protein control fragment (Product #RP-107737). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction