missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR alpha/NR1C1 Antibody (3B6/PPAR), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB300-537APC
This item is not returnable.
View return policy
Description
PPAR alpha/NR1C1 Monoclonal specifically detects PPAR alpha/NR1C1 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Flow Cytometry.
Specifications
| PPAR alpha/NR1C1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| PPARA | |
| Purified recombinant PPAR alpha protein. | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Transcription Factors and Regulators | |
| 5465 | |
| Human, Mouse, Rat, Bovine, Rabbit | |
| Purified |
| Flow Cytometry | |
| 3B6/PPAR | |
| Flow Cytometry | |
| alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| PPAR alpha (3B6/PPAR) | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction