missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR alpha/NR1C1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 302.00 - € 498.00
Specifications
| Antigen | PPAR alpha/NR1C1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18232401
|
Novus Biologicals
NBP2-58507 |
100 μL |
€ 498.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610658
|
Novus Biologicals
NBP2-58507-25ul |
25 μL |
€ 302.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPAR alpha/NR1C1 Polyclonal specifically detects PPAR alpha/NR1C1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| PPAR alpha/NR1C1 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Lipid and Metabolism, mTOR Pathway, Transcription Factors and Regulators | |
| alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452 | |
| PPARA | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 5465 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FGRMPRSEKAKLKAEILTCEHDIEDSETADLKSLAKRIYEAYLKN | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title